
Spirodiclofen

Synonym(s):
3-(2,4-Dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl) 2,2-dimethylbutyrate
Empirical Formula (Hill Notation):
C21H24Cl2O4
CAS Number:
Molecular Weight:
411.32
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2