
Sodium molybdate dihydrate

Synonym(s):
Molybdic acid sodium salt dihydrate, Sodium molybdate dihydrate
Linear Formula:
Na2MoO4 · 2H2O
CAS Number:
Molecular Weight:
241.95
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing