Skip to Content
Merck
Sodium dimethyldithiocarbamate hydrate 98%

Sodium dimethyldithiocarbamate hydrate

Sodium dimethyldithiocarbamate hydrate 98%
Synonym(s):
Dimethyldithiocarbamic acid sodium salt
Linear Formula:
(CH3)2NCSSNa · aq
CAS Number:
Molecular Weight:
143.21 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing