
Sodium 2-mercaptoethanesulfonate

Synonym(s):
2-Mercaptoethanesulfonic acid sodium salt, Coenzyme M sodium salt, HS-CoM Na, MESNA
Linear Formula:
HSCH2CH2SO3Na
CAS Number:
Molecular Weight:
164.18
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing