Skip to Content
Merck
Silver acetate 99.99% trace metals basis

Silver acetate

Silver acetate 99.99% trace metals basis
Synonym(s):
Acetic acid silver salt
Linear Formula:
CH3COOAg
CAS Number:
Molecular Weight:
166.91
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing