Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Selenium dioxide
Synonym(s):
NSC 56753, Selenium oxide
Linear Formula:
SeO
2
CAS Number:
7446-08-4
Molecular Weight:
110.96
EC Number:
231-194-7
Products
Brand
Sigma-Aldrich (4)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
325473
Selenium dioxide,
reagent grade, powder, 98%
Expand
View Pricing
200107
Selenium dioxide,
≥99.9% trace metals basis
Expand
View Pricing
204315
Selenium dioxide,
99.999% trace metals basis
Expand
View Pricing
213365
Selenium dioxide,
ReagentPlus
®
, powder, 99.8% trace metals basis
Expand
View Pricing