
Selenium

Synonym(s):
Selenium
Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Element
Quality Segment
Showing 1-10 of 10
Compare
Product Number
Description
Pricing