Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rhodamine B isothiocyanate
Synonym(s):
RBITC
Empirical Formula (Hill Notation):
C
29
H
30
ClN
3
O
3
S
CAS Number:
36877-69-7
Molecular Weight:
536.08
EC Number:
253-248-9
Products
Product Category
dyes (1)
stains (1)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Physical Form
Product Line
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
R1755
Rhodamine B isothiocyanate,
mixed isomers, BioReagent, suitable for protein labeling
Expand
View Pricing
283924
Rhodamine B isothiocyanate,
mixed isomers
Expand
View Pricing