
Ramipril Related Compound A

Synonym(s):
(2S,3aS,6aS)-1-{(2S)-2-{[(1S)-1-(Methoxycarbonyl)-3-phenylpropyl]amino}-1-oxopropyl}octahydrocyclopenta[b]pyrrole-2-carboxylic acid, Ramipril methyl ester
Empirical Formula (Hill Notation):
C22H30N2O5
CAS Number:
Molecular Weight:
402.48
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2