Skip to Content
Merck
Pyrocatechol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Pyrocatechol

Pyrocatechol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
1,2-Benzenediol, 1,2-Dihydroxybenzene, 2-Hydroxyphenol, Catechol
Linear Formula:
C6H4-1,2-(OH)2
CAS Number:
Molecular Weight:
110.11
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Color
Reference Material Type
Special Grade
Technique