Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
hollow stopper
Synonym(s):
Corning
®
7650
Products
Product Category
stoppers (6)
Manufacturer
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS76508
Pyrex
®
hollow stopper,
stopper joint: 8
Expand
View Pricing
CLS765022
Pyrex
®
hollow stopper,
stopper joint: 22
Expand
View Pricing
CLS765016
Pyrex
®
hollow stopper,
stopper joint: 16
Expand
View Pricing
CLS765013
Pyrex
®
hollow stopper,
stopper joint: 13
Expand
View Pricing
CLS76509
Pyrex
®
hollow stopper,
stopper joint: 9
Expand
View Pricing
CLS765019
Pyrex
®
hollow stopper,
stopper joint: 19
Expand
View Pricing