Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PTFE Membrane Filter, 5.0 μm Pore Size
Products
Product Category
filter discs (3)
Brand
Millipore (5)
Manufacturer
Agency
Diameter (mm)
Quality Segment
Hydrophobicity
Color
Membrane Material
Pore size (µm)
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
JMWP02500
PTFE Membrane Filter, 5.0 μm Pore Size,
Omnipore, filter diam. 25 mm, hydrophilic
Expand
View Pricing
JMWP14225
PTFE Membrane Filter, 5.0 μm Pore Size,
Omnipore, filter diam. 142 mm, hydrophilic
Expand
View Pricing
FMLW02500
PTFE Membrane Filter, 5.0 μm Pore Size,
Fluoropore
®
, filter diam. 25 mm, hydrophobic
Expand
View Pricing
LSWP03700
PTFE Membrane Filter, 5.0 μm Pore Size,
Mitex
™
, filter diam. 37 mm, hydrophobic
Expand
View Pricing
FMLW03700
PTFE Membrane Filter, 5.0 μm Pore Size,
Fluoropore
®
, filter diam. 37 mm, hydrophobic
Expand
View Pricing