Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Prostaglandin E
2
Synonym(s):
(5Z,11α,13E,15S)-11,15-Dihydroxy-9-oxoprosta-5,13-dienoic acid, Dinoprostone, PGE
2
Empirical Formula (Hill Notation):
C
20
H
32
O
5
CAS Number:
363-24-6
Molecular Weight:
352.47
EC Number:
206-656-6
Products
Product Category
hormones (2)
lipids (1)
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
P0409
Prostaglandin E
2
,
synthetic, powder, BioReagent, suitable for cell culture
Expand
View Pricing
P5640
Prostaglandin E
2
,
≥93% (HPLC), synthetic
Expand
View Pricing
P6532
Prostaglandin E
2
,
γ-irradiated, powder, BioXtra, suitable for cell culture
Expand
View Pricing