Skip to Content
Merck
Propyl 4-hydroxybenzoate sodium salt tested according to Ph. Eur.

Propyl 4-hydroxybenzoate sodium salt

Propyl 4-hydroxybenzoate sodium salt tested according to Ph. Eur.
Synonym(s):
Sodium propyl 4-hydroxybenzoate
Empirical Formula (Hill Notation):
C10H11NaO3
CAS Number:
Molecular Weight:
202.18
EC Number:

Products

Product Category
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Special Grade
Showing 1-2 of 2