
Potassium chloride solution

Synonym(s):
Potassium chloride solution, Potassium chloride solution saturated with AgCl
Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Format
Quality Segment
Application
Reaction Type
Reference Material Type
Special Grade
Technique
Showing 1-12 of 12
Compare
Product Number
Description
Pricing