Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium chloride
Synonym(s):
Potassium chloride
Linear Formula:
KCl
CAS Number:
7447-40-7
Molecular Weight:
74.55
EC Number:
231-211-8
Products
Product Category
salts (17)
Brand
Sigma-Aldrich (15)
Supelco (4)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
Anhydrous (3)
ACS reagent (2)
Puriss (2)
Puriss P.A. (1)
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
P3911
Potassium chloride,
ACS reagent, 99.0-100.5%
Expand
View Pricing
1.04936
Potassium chloride,
for analysis EMSURE
®
Expand
View Pricing
12636
Potassium chloride,
puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Expand
View Pricing
P9541
Potassium chloride,
for molecular biology, ≥99.0%
Expand
View Pricing
P5405
Potassium chloride,
powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Expand
View Pricing
P4504
Potassium chloride,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
746436
Potassium chloride,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
60130
Potassium chloride,
puriss. p.a., ≥99.5% (AT)
Expand
View Pricing
60128
Potassium chloride,
BioUltra, for molecular biology, ≥99.5% (AT)
Expand
View Pricing
1.04933
Potassium chloride,
for analysis (<= 0.005% Br) EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
1.04938
Potassium chloride,
99.999 Suprapur
®
Expand
View Pricing
793590
Potassium chloride,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99%
Expand
View Pricing
409316
Potassium chloride,
99.999% trace metals basis
Expand
View Pricing
204099
Potassium chloride,
≥99.99% trace metals basis
Expand
View Pricing
1548190
Potassium chloride,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
P9333
Potassium chloride,
BioXtra, ≥99.0%
Expand
View Pricing
PHR1329
Potassium chloride,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
449989
Potassium chloride,
AnhydroBeads
™
, −10 mesh, 99.999% trace metals basis
Expand
View Pricing
451029
Potassium chloride,
AnhydroBeads
™
, −10 mesh, 99.99% trace metals basis
Expand
View Pricing
NIST999C
Potassium chloride,
Primary Standard, NIST
®
SRM
®
Expand
View Pricing
Page 1 of 2
Page 1 of 2