
Potassium antimony(III) tartrate hydrate

Synonym(s):
Antimony potassium tartrate
Empirical Formula (Hill Notation):
C8H4K2O12Sb2 · xH2O
CAS Number:
Molecular Weight:
613.83 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing