Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyvinylpyrrolidone
Synonym(s):
PVP, Polyvidone, Povidone
Linear Formula:
(C
6
H
9
NO)
n
CAS Number:
9003-39-8
Products
Product Category
detergents (3)
Brand
Sigma-Aldrich (13)
Biological Source
Feature
Physical Form
Product Line
Quality Segment
Application
Polymer Composition
Polymer Type
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
PVP360
Polyvinylpyrrolidone,
mol wt (number average molecular weight Mn 360kDa)
Expand
View Pricing
PVP40
Polyvinylpyrrolidone,
average mol wt 40,000
Expand
View Pricing
437190
Polyvinylpyrrolidone,
average M
w
~1,300,000 by LS
Expand
View Pricing
856568
Polyvinylpyrrolidone,
powder, average M
w
~55,000
Expand
View Pricing
234257
Polyvinylpyrrolidone,
powder, average M
w
~29,000
Expand
View Pricing
P0930
Polyvinylpyrrolidone,
powder, BioXtra, suitable for mouse embryo cell culture
Expand
View Pricing
PVP10
Polyvinylpyrrolidone,
average mol wt 10,000
Expand
View Pricing
81420
Polyvinylpyrrolidone,
K 30
Expand
View Pricing
81440
Polyvinylpyrrolidone,
K 90
Expand
View Pricing
P5288
Polyvinylpyrrolidone,
for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Expand
View Pricing
90268
Polyvinylpyrrolidone,
K 25, tested according to Ph. Eur.
Expand
View Pricing
P2307
Polyvinylpyrrolidone,
suitable for plant cell culture, average mol wt 10,000
Expand
View Pricing
RDD033
Polyvinylpyrrolidone,
free-flowing, Redi-Dri
™
, powder, average mol wt 40,000
Expand
View Pricing