
Plastic additive 27

Synonym(s):
Dioctyl terephthalate, Bis(2-ethylhexyl) terephthalate
Linear Formula:
C6H4-1,4-[CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
390.56
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing