
Perfluorobutanoic acid

Synonym(s):
Heptafluorobutyric acid, Edman Reagent No. 3, HFBA, Perfluorobutyric acid
Linear Formula:
CF3CF2CF2COOH
CAS Number:
Molecular Weight:
214.04
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing