Pentamethylcyclopentadienylrhodium(III) chloride dimer
Synonym(s):
1,3-Cyclopentadiene, 1,2,3,4,5-pentamethyl-, rhodium complex, Bis(dichloro(pentamethylcyclopentadienyl)rhodium), Bis(pentamethylcyclopentadienylrhodium dichloride), Dichloro(pentamethylcyclopentadienyl)rhodium(II) dimer, Tetrachlorobis(pentamethylcyclopentadienyl)dirhodium
Linear Formula:
[Rh(C5Me5)Cl2]2
CAS Number:
Molecular Weight:
618.08
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing