Skip to Content
Merck
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 97%

N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate

N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 97%
Synonym(s):
O-(N-Succinimidyl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate, TSTU
Empirical Formula (Hill Notation):
C9H16BF4N3O3
CAS Number:
Molecular Weight:
301.05

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1