Skip to Content
Merck
N,N-Dimethylethylenediamine ≥98.0% (GC)

N,N-Dimethylethylenediamine

N,N-Dimethylethylenediamine ≥98.0% (GC)
Synonym(s):
2-(Dimethylamino)ethylamine, DMEN
Linear Formula:
(CH3)2NCH2CH2NH2
CAS Number:
Molecular Weight:
88.15
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing