Skip to Content
Merck
Nimodipine European Pharmacopoeia (EP) Reference Standard

Nimodipine

Nimodipine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-methoxyethyl 1-methylethyl ester, Isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-(m-nitrophenyl)-3,5-pyridinedicarboxylate, Nimodipine
Empirical Formula (Hill Notation):
C21H26N2O7
CAS Number:
Molecular Weight:
418.44

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique