
Nicotinamide Impurity E

Synonym(s):
Nicotinamide N-oxide
Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing