N-Glycolylneuraminic acid
Synonym(s):
Neu5Glc, NeuNGl
Empirical Formula (Hill Notation):
C11H19NO10
CAS Number:
Molecular Weight:
325.27
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing