
Naphazoline impurity A

Synonym(s):
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide
Empirical Formula (Hill Notation):
C14H16N2O
CAS Number:
Molecular Weight:
228.29
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing