Mevastatin
Synonym(s):
Compactin, ML-236B, Mevastatin
Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing