
Metoclopramide

Synonym(s):
4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide, Methoxychloroprocainamide
Empirical Formula (Hill Notation):
C14H22ClN3O2
CAS Number:
Molecular Weight:
299.80
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing