Skip to Content
Merck
Methyl Red hydrochloride ACS reagent

Methyl Red hydrochloride

Methyl Red hydrochloride ACS reagent
Synonym(s):
4-Dimethylaminoazobenzene-2′-carboxylic acid hydrochloride, Acid Red 2
Linear Formula:
(CH3)2NC6H4N=NC6H4CO2H·HCl
CAS Number:
Molecular Weight:
305.76
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing