
Methyl myristelaidate

Synonym(s):
Methyl trans-9-tetradecenoate, Myristelaidic acid methyl ester
Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing