
Methylammonium iodide

Synonym(s):
Methanamine hydriodide
Linear Formula:
CH3NH2 • HI
CAS Number:
Molecular Weight:
158.97
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing