
Mesityl oxide

Synonym(s):
4-Methyl-3-penten-2-one, Mesityl oxide
Linear Formula:
(CH3)2C=CHCOCH3
CAS Number:
Molecular Weight:
98.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3