
Medronic acid

Synonym(s):
Methylenediphosphonic acid, MDP, Medronic acid
Linear Formula:
CH2[P(O)(OH)2]2
CAS Number:
Molecular Weight:
176.00
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing