![Magnesium Chloride, Hexahydrate, Molecular Biology Grade](/deepweb/assets/sigmaaldrich/product/structures/653/744/67271700-7b55-4e0c-84c7-4b6e02bb58c1/640/67271700-7b55-4e0c-84c7-4b6e02bb58c1.png)
Magnesium Chloride, Hexahydrate, Molecular Biology Grade
![Magnesium Chloride, Hexahydrate, Molecular Biology Grade](/deepweb/assets/sigmaaldrich/product/structures/653/744/67271700-7b55-4e0c-84c7-4b6e02bb58c1/640/67271700-7b55-4e0c-84c7-4b6e02bb58c1.png)
Synonym(s):
Magnesium chloride hexahydrate
Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing