Macrophage Colony-Stimulating Factor human
Synonym(s):
MCSF
Products
Product Category
Brand
Biological Source
Color
Gene Alias
Manufacturer
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing