Skip to Content
Merck
Lucifer Yellow VS dilithium salt ~85%

Lucifer Yellow VS dilithium salt

Lucifer Yellow VS dilithium salt ~85%
Synonym(s):
4-Amino-N-(3-[vinylsulfonyl]phenyl)naphthalimide-3,6-disulfonate dilithium salt, Dilithium 4-amino-N-[3-(vinylsulfonyl)phenyl]naphthalimide-3,6-disulfonate
Empirical Formula (Hill Notation):
C20H12Li2N2O10S3
CAS Number:
Molecular Weight:
550.39

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing