Skip to Content
Merck
Lithium iodide AnhydroBeads™, −10 mesh, 99.99% trace metals basis

Lithium iodide

Lithium iodide AnhydroBeads™, −10 mesh, 99.99% trace metals basis
Linear Formula:
LiI
CAS Number:
Molecular Weight:
133.85
EC Number:

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Special Grade