Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium fluoride
Synonym(s):
Fluorolithium
Linear Formula:
LiF
CAS Number:
7789-24-4
Molecular Weight:
25.94
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (5)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Particle Size (µm)
Special Grade
Anhydrous (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
237965
Lithium fluoride,
powder, -300 mesh
Expand
View Pricing
449903
Lithium fluoride,
≥99.99% trace metals basis
Expand
View Pricing
203645
Lithium fluoride,
powder, <100 μm, ≥99.98% trace metals basis
Expand
View Pricing
669431
Lithium fluoride,
Precipitated, 99.995%
Expand
View Pricing
932310
Lithium fluoride,
Expand
View Pricing