
Lanthanum(III) trifluoromethanesulfonate

Synonym(s):
Lanthanum(III) triflate, Trifluoromethanesulfonic acid lanthanum salt
Linear Formula:
(CF3SO3)3La
CAS Number:
Molecular Weight:
586.11
Products
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing