
Isobutylmagnesium chloride solution

Linear Formula:
(CH3)2CHCH2MgCl
CAS Number:
Molecular Weight:
116.87
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing