Skip to Content
Merck
Inosine 5′-triphosphate trisodium salt ≥95%

Inosine 5′-triphosphate trisodium salt

Inosine 5′-triphosphate trisodium salt ≥95%
Synonym(s):
ITP
Empirical Formula (Hill Notation):
C10H12Na3N4O14P3
CAS Number:
Molecular Weight:
574.11

Products

Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing