Skip to Content
Merck
Germanium microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%

Germanium

Germanium microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Synonym(s):
Germanium
Empirical Formula (Hill Notation):
Ge
CAS Number:
Molecular Weight:
72.64

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Diameter (mm)
Length (mm)
Element
Quality Segment
Showing 1-20 of 33
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2