Skip to Content
Merck
Furfuryl alcohol analytical standard

Furfuryl alcohol

Furfuryl alcohol analytical standard
Synonym(s):
2-(Hydroxymethyl)furan
Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing