
Fenamidone

Synonym(s):
(S)-1-Anilino-4-methyl-2-methylthio-4-phenyl-2-imidazolin-5-one
Empirical Formula (Hill Notation):
C17H17N3OS
CAS Number:
Molecular Weight:
311.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2