Skip to Content
Merck
Ethyl palmitate analytical standard

Ethyl palmitate

Ethyl palmitate analytical standard
Synonym(s):
Ethyl hexadecanoate, Palmitic acid ethyl ester
Linear Formula:
CH3(CH2)14COOC2H5
CAS Number:
Molecular Weight:
284.48
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4