Skip to Content
Merck
Ethanolamine ReagentPlus®, ≥99%

Ethanolamine

Ethanolamine ReagentPlus®, ≥99%
Synonym(s):
2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine
Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique