Skip to Content
Merck
Esfenvalerate PESTANAL®, analytical standard

Esfenvalerate

Esfenvalerate PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C25H22ClNO3
CAS Number:
Molecular Weight:
419.90

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing