
Eleutheroside B

Synonym(s):
(E)-4-[-3-Hydroxy-1-propenyl]-2,6-dimethoxyphenyl-β-D-glucopyranoside, Ilexanthin A, Ligustrin, Lilacin, Magnolenin, Methoxyconiferine, Sinapyl alcohol 4-O-glucoside, Syringin, Syringoside
Empirical Formula (Hill Notation):
C17H24O9
CAS Number:
Molecular Weight:
372.37
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3