
Dopamine-1,1,2,2-d4 hydrochloride

Synonym(s):
2-(3,4-Dihydroxyphenyl)ethyl-1,1,2,2-d4-amine hydrochloride
Linear Formula:
C8D4H7O2N.HCl
CAS Number:
Molecular Weight:
193.66
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing